Läs mer
Sodium 2-Hydrazinylacetate, TRC
Produktkod. 30420642
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30420642 1 g
1 options
Produktkod. 30420642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS5979151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium 2-Hydrazinylacetate
CAS 94107-53-6
Molekylformel C2H5N2NaO2
Kvantitet 1 g
Synonym Acetic acid','2-hydrazinyl-','sodium salt (1:1), Acetic acid','hydrazino-','monosodium salt (9CI), Sodium hydrazinoacetate, Hydrazinoacetic acid sodium salt, Sodium 2-hydrazinylacetate
InChI formel InChI=1S/C2H6N2O2.Na/c3-4-1-2(5)6;/h4H,1,3H2,(H,5,6);/q;+1/p-1
LEDER [Na+].NNCC(=O)[O-]
IUPAC-namn sodium;2-hydrazinylacetate
Molekylvikt (g/mol) 112.06
Formel vikt 112.0249