Läs mer
Sodium 2-Nitrophenolate, TRC
Produktkod. 30011801
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011801 2.5 g 2.5g
30009319 250 mg -
30009678 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011801 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6343152.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 824-39-5
Molekylformel C6 H4 N O3 . Na
Molekylvikt (g/mol) 161.09
Synonym Phenol, 2-nitro-, sodium salt (1:1), Phenol, 2-nitro-, sodium salt (9CI), Phenol, o-nitro-, sodium deriv. (6CI), Phenol, o-nitro-, sodium salt (8CI), Sodium, (o-nitrophenoxy)- (7CI), (o-Nitrophenolato)sodium, 2-Nitrophenol sodium salt, Sodium 2-nitrophenolate, Sodium 2-nitrophenoxide, Sodium o-nitrophenol, Sodium o-nitrophenolate, Sodium o-nitrophenoxide, o-Nitrophenol sodium salt
IUPAC-namn sodium;2-nitrophenolate
LEDER [Na+].[O-]c1ccccc1[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium 2-Nitrophenolate
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C6H5NO3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H;/q;+1/p-1
Formel vikt 161.0089
Procent renhet >95%