Läs mer
Sodium 4-phenylbutyrate, MedChemExpress
Produktkod. 16471186
Encompass_Preferred
Change view
Kvantitet:
100 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16471186 200 mg
16426866 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16471186 Leverantör Medchem Express Leverantörsnummer HY15654
MedChemExpress Sodium 4-phenylbutyrate (4-PBA sodium) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research.
Sodium 4-phenylbutyrate (4-PBA sodium) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research.
Specifikationer
Kemiskt namn eller material Sodium 4-phenylbutyrate
Fysisk form Powder
CAS 1716-12-7
Färg White
Kvantitet 200 mg
Anteckningar om renhetsgrad Research
Molekylformel C10H11NaO2
För användning med (applikation) Cancer-programmed cell death
Synonym 4-PBA sodium4-Phenylbutyric acid sodiumBenzenebutyric acid sodium
Löslighetsinformation H2O : 23.5 mg/mL (126.22 mM; Need ultrasonic and warming)
LEDER O=C(O[Na])CCCC1=CC=CC=C1
Molekylvikt (g/mol) 186.18
Formel vikt 186.18
Procent renhet 98.55%