Läs mer
Sodium Acetate-1,2-13C2, TRC
Produktkod. 30136610
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136610 1 g
30135852 50 mg
30006711 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6109711G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56374-56-2
Molekylformel 13C2H3NaO2
Molekylvikt (g/mol) 84.02
Synonym Sodium [13C2]-Acetate, Acetic-13C2 Acid Sodium Salt
IUPAC-namn sodium;acetate
LEDER [Na+].[13CH3][13C](=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium Acetate-1,2-13C2
Kvantitet 1 g
InChI formel InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1,2+1;
Formel vikt 84.0098