Läs mer
Sodium Acetate (2-13C), TRC
Produktkod. 30134049
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134049 100 mg
30135433 10 mg
17937254 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS960290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13291-89-9
Molekylformel C13CH3NaO2
Molekylvikt (g/mol) 83.03
Synonym Sodium [2-13C]acetate, Sodium Acetate-2-13C, [2-13C]Sodium Acetate
IUPAC-namn sodium;acetate
LEDER [Na+].[13CH3]C(=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium Acetate (2-13C)
Kvantitet 100 mg
InChI formel InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1;
Formel vikt 83.0064