missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 7.0, RNAse free, Thermo Scientific Chemicals

Catalog Number p-7042269
Click to view available options
Quantity:
125 mL
250 mL
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O

Sodium acetate buffer solution pH, 7.0 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium acetate buffer solution pH, 7.0 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .
TRUSTED_SUSTAINABILITY
pH 7.0
Physical Form Liquid
Quantity 125 mL
Linear Formula NaOOCCH3
IUPAC Name sodium acetate
Formula Weight 82.03
Concentration or Composition (by Analyte or Components) 3M aq. soln.
Chemical Name or Material Sodium acetate, RNAse free

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'