Läs mer
Sodium Acrylate, TRC
Produktkod. 30412978
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30412978 5 g
1 options
Produktkod. 30412978 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS9600605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium Acrylate
CAS 7446-81-3
Molekylformel C3 H3 O2 . Na
Kvantitet 5 g
Synonym 2-Propenoic acid','sodium salt (1:1) (ACI), 2-Propenoic acid','sodium salt (9CI), Acrylic acid','sodium salt (6CI','7CI','8CI), CI 05K, Flexisperse 450N, Longhai 1240, Sodium 2-propenoate, Sodium acrylate, Webac 240A1
InChI formel InChI=1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1
LEDER [Na+].[O-]C(=O)C=C
IUPAC-namn sodium;prop-2-enoate
Molekylvikt (g/mol) 94.044
Formel vikt 94.003