missing translation for 'onlineSavingsMsg'
Learn More

Sodium Aluminate, TRC

Product Code. 30410370
Change view
Click to view available options
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30410370 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410370 Supplier Toronto Research Chemicals Supplier No. TRCS22449010G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Sodium Aluminate
CAS 11138-49-1
Molekylformel NaAlO2
Kvantitet 10 g
Synonym Dynaflock L, J 242, Maxifloc 8010, Monofrax H, NA 150, NA 150 (oxide), NA 170, Nalco 680, Sodium Aluminate, Sodium Aluminum Oxide, Sodium Beta Alumina, Sodium Polyaluminate, Sodium β-Alumina, Sodium β''-Alumina, Torganit, VSA 45, β-Alumina
InChI formel InChI=1S/Al.Na.2O/q-1;+1;;
LEDER O=[Al-]=O.[Na+]
Molekylvikt (g/mol) 81.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.