Learn More
Sodium Azide, Sigma-Aldrich™
Product Code. 15826760
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15826760 100 g 100g
15891801 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15826760 Supplier Sigma Aldrich Supplier No. 8223350100
 Restricted Item
Beräknad leverans: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26628-22-8
Specifications
Boiling Point 300°C/1013 hPa (rigorous decomposition)
Melting Point 275°C (decomposition)
Quantity 100 g
Density 1.85 g/cm3 at 20°C
MDL Number MFCD00003536
Synonym Sodium azide, Hydrazoic azid sodium salt, Sodium azide
Molecular Weight (g/mol) 65.01
Grade Research
Chemical Name or Material Sodium Azide