Läs mer
Sodium Benzenethiolate (technical grade, 90%), TRC
Produktkod. 30428156
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428156 500 mg
30411841 5 g
30422431 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428156 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS930690500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 930-69-8
Molekylformel C6H5NaS
Molekylvikt (g/mol) 132.16
Synonym Sodium Phenylmercaptide, Sodium Phenylsulfide, Sodium Phenylthiolate, Sodium Thiophenate, Sodium Thiophenolate, Sodium Thiophenoxide, Sodium Thiophenylate, Thiophenol Sodium Salt, Thiophenyl Sodium Salt
IUPAC-namn sodium;benzenethiolate
LEDER [Na+].[S-]c1ccccc1
Specifikationer
Kemiskt namn eller material Sodium Benzenethiolate (technical grade)
Kvantitet 500 mg
InChI formel InChI=1S/C6H6S.Na/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1
Formel vikt 132.001