missing translation for 'onlineSavingsMsg'
Läs mer

Sodium Benzoate-d5, TRC

Produktkod. 30412474
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412474 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412474 Supplier Toronto Research Chemicals Supplier No. TRCS614001100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Sodium Benzoate-d5
CAS 62790-26-5
Molekylformel C7 D5 O2 . Na
Kvantitet 100 mg
Synonym Benzoic-d5 acid','sodium salt (9CI), Sodium benzoate-d5, Benzoic-2,3,4,5,6-d5 acid sodium salt, 2,3,4,5,6-Pentadeuteriobenzoic acid sodium salt, Benzoic Acid-D5 (phenyl-D5) Sodium Salt
InChI formel InChI=1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1/i1D,2D,3D,4D,5D;
LEDER [Na+].[2H]c1c([2H])c([2H])c(C(=O)[O-])c([2H])c1[2H]
IUPAC-namn sodium;2,3,4,5,6-pentadeuteriobenzoate
Molekylvikt (g/mol) 149.134
Formel vikt 149.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.