Läs mer
Sodium beta-Hydroxyethoxyacetate, TRC
Produktkod. 30412619
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412619 1 g
30409754 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6348501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142047-97-0
Molekylformel C4H7NaO4
Molekylvikt (g/mol) 142.09
Synonym (2-Hydroxyethoxy)acetic Acid Monosodium Salt, Sodium β-Hydroxyethoxyacetate
LEDER O=C(COCCO)O[Na]
Specifikationer
Kemiskt namn eller material Sodium beta-Hydroxyethoxyacetate
Kvantitet 1 g
InChI formel InChI=1S/C4H8O4.Na/c5-1-2-8-3-4(6)7;/h5H,1-3H2,(H,6,7);/q;+1/p-1
Formel vikt 142.0242