missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, for biochemistry, Thermo Scientific Chemicals

Catalog Number p-4813646
Click to view available options
Quantity:
500 g
2.5 kg
6132-02-1
CNa2O3·10H2O
286.14
XYQRXRFVKUPBQN-UHFFFAOYSA-L
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
151402
disodium;carbonate;decahydrate
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
6132-02-1
CNa2O3·10H2O
286.14
XYQRXRFVKUPBQN-UHFFFAOYSA-L
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
151402
disodium;carbonate;decahydrate
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34.0°C
Quantity 500 g
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Merck Index 15,8731
Solubility Information Clear colorless
Formula Weight 286.14
Percent Purity ≥99%
Grade Biochemical
Specific Gravity 1.46
Density 1.4600g/mL
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate decahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'