Learn More
Sodium Carbonate, Supelco™
Product Code. 15824810
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824810 1 kg 1kg
15814810 500 g 500g
15807460 5 kg 5kg
15817460 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15824810 Supplier Supelco Supplier No. 1063921000
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
I lager
Add to basket
Edge
Add to basket
Anhydrous for analysis EMSURE™ ISO
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 497-19-8
Specifications
Melting Point 851°C (lit.)
Quantity 1 kg
Molecular Formula Na2CO3
MDL Number MFCD00003494
Synonym anhydrous soda, Calcined soda, Carbonic acid disodium salt, Soda ash, Calcined soda, Carbonic acid disodium salt, Soda ash
Molecular Weight (g/mol) 105.99
Grade Research
Density 2.53 g/cm3 at 20°C
Chemical Name or Material Sodium Carbonate
compliance-icons
Product Identifier
  • Sodium carbonate anhydrous for analysis EMSURE® ISO
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.