Learn More
Sodium Chloride, ACS, 99.5% min, MilliporeSigma™
Product Code. 15864810
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15864810 1 kg Plastic Bottle 1kg
15854810 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864810 Supplier Supelco Supplier No. 1064041000
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
I lager
Add to basket
Edge
Add to basket
Chemical Identifiers
CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
Specifications
Color Colorless
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99.5% min.
Formula Weight 58.44
Percent Purity ≥99.5%
Grade ACS
Packaging Plastic Bottle
Vapor Pressure 1.3 hPa (865°C)
Density 2.17 g/cm3 (20°C)
Chemical Name or Material Sodium Chloride, For ACS analysis