Learn More
Alfa Aesar™ Sodium chloride crystal optic disc, 13mm x 1mm, polished both sides
Spectroscopy, IR window
Brand:  Alfa Aesar™ 038443.KT
Additional Details : Weight : 0.01000kg
Product Code. 15466235
  • 399.16 SEK / Each
EU Stock 2
Add to basket
Specifications
Specifications
Sodium chloride crystal optic disc
Odorless
1 each
13mm x 1mm, Polished both sides