missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride USP GenAR Granular, Macron Fine Chemicals™

Catalog Number 15518434
Click to view available options
POLYSTORMOR; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 7647-14-5
Physical Form Granules
Quantity 500 g
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Grade USP
Packaging POLYSTORMOR
Chemical Name or Material Sodium Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'