missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium citrate, 99.8%, For ACS analysis, MP Biomedicals™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Color | White |
Quantity | 500 g |
Formula Weight | 294.1 |
Percent Purity | 0.998 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Sodium citrate, For ACS analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'