missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals

Catalog Number p-4407879
Click to view available options
Quantity:
5 g
25 g
100 g
25895-60-7
CH3BNNa
62.84
MFCD00003516
CVDUGUOQTVTBJH-UHFFFAOYSA-N
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
20587905
[Na+].[BH3-]C#N
25895-60-7
CH3BNNa
62.84
MFCD00003516
CVDUGUOQTVTBJH-UHFFFAOYSA-N
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
20587905
[Na+].[BH3-]C#N

Selective Reducing Agent For Aldehydes, Ketones, Oximes

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
TRUSTED_SUSTAINABILITY
Melting Point >300°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 95%
Solubility Information Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether.
IUPAC Name sodium;cyanoboron(1-)
Formula Weight 62.84
Sensitivity Moisture sensitive
Density 1.20 g/cm3 at 20°C
Chemical Name or Material Sodium cyanoborohydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'