Läs mer
Sodium Dibromomethanesulfonate, TRC
Produktkod. 17911475
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17911475 10 mg
17972485 50 mg
17964695 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17911475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS62411010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 859073-88-4
Molekylformel CHBr2NaO3S
Molekylvikt (g/mol) 275.88
Synonym 1,1-Dibromomethanesulfonic Acid Sodium Salt
IUPAC-namn sodium;dibromomethanesulfonate
LEDER [Na+].[O-]S(=O)(=O)C(Br)Br
Specifikationer
Kemiskt namn eller material Sodium Dibromomethanesulfonate
Kvantitet 10 mg
InChI formel InChI=1S/CH2Br2O3S.Na/c2-1(3)7(4,5)6;/h1H,(H,4,5,6);/q;+1/p-1
Formel vikt 273.7911