Läs mer
Sodium Dichloromethanesulfonate, TRC
Produktkod. 30410592
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30410592 50 mg
1 options
Produktkod. 30410592 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS62410550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium Dichloromethanesulfonate
CAS 53638-47-4
Molekylformel CHCl2NaO3S
Kvantitet 50 mg
Synonym 1,1 -Dichloro-methanesulfonic Acid Sodium Salt
InChI formel InChI=1S/CH2Cl2O3S.Na/c2-1(3)7(4,5)6;/h1H,(H,4,5,6);/q;+1/p-1
LEDER [Na+].[O-]S(=O)(=O)C(Cl)Cl
IUPAC-namn sodium;dichloromethanesulfonate
Molekylvikt (g/mol) 186.98
Formel vikt 185.8921