Learn More
Sodium Dihydrogen Phosphate Monohydrate, EMSURE™, MilliporeSigma™
Product Code. 15857450
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Carton
Unit Size:
25kg
Product Code. Quantity Packaging Unit Size
15857450 25 kg Fiber Carton 25kg
1 options
Product Code. 15857450 Supplier Supelco Supplier No. 1063469025
 Restricted Item
Beräknad leverans: 20-11-2026
Chemical Identifiers
CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O
Specifications
Physical Form Crystals
Quantity 25 kg
Percent Purity 99.0 to 102.0%
Grade ACS
Packaging Fiber Carton
Density 2.04 g/3 (20 °C)
Chemical Name or Material Sodium Dihydrogen Phosphate Monohydrate