missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Estrone-2,4,16,16-d4 Sulfate (Stabilized with TRIS, 50% w/w), CDN
Specifications
Specifications
| Kemiskt namn eller material | Estrone-2,4,16,16-d4 3-Sulfate sodium salt |
| CAS | 285979-80-8 |
| Lösningsmedel eller matris | TRIS |
| Molekylformel | C18 2H4 H17 O5 S . Na |
| Kvantitet | 0.1 g |
| Synonym | Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt (1:1), Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt (9 CI), Estrone-2,4,16,16-D4 3-sulfate sodium |
| InChI formel | InChI=1 S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-16 H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,18+;/m1./s1/i3D,7D2,10 D; |
| LEDER | [Na+].[2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)[C@@H]3 CCc2c([2 H])c1OS(=O)(=O)[O-] |
| IUPAC-namn | sodium;[(8 R,9 S,13 S,14 S)-2,4,16,16-tetradeuterio-13-methyl-17-oxo-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-yl] sulfate |
| Molekylvikt (g/mol) | 376.44 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?