Läs mer
Sodium Ethyl Parahydroxybenzoate, LoGiCal
Produktkod. 30462352
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462352 10 mg
1 options
Produktkod. 30462352 Leverantör LoGiCal Leverantörsnummer LGCFOR0498.05
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium Ethyl Parahydroxybenzoate
CAS 35285-68-8
Analyt- eller komponentnamn Sodium Ethyl Parahydroxybenzoate
Frakt skick Room Temperature
Molekylformel C9 H9 O3 . Na
Certifikat och efterlevnad ISO 9001
Synonym Sodium Ethyl Parahydroxybenzoate, p-Hydroxybenzoic acid ethyl ester sodium deriv., 4-Hydroxybenzoic acid ethyl ester sodium salt, Ethyl 4-hydroxybenzoate sodium salt, Ethyl p-hydroxybenzoate sodium salt, Sodium ethyl p-hydroxybenzoate, Sodium p-(carbethoxy)phenoxide
InChI formel InChI=1S/C9H10O3.Na/c1-2-12-9(11)7-3-5-8(10)6-4-7;/h3-6,10H,2H2,1H3;/q;+1/p-1
Lösningstyp Neat
LEDER [Na+].CCOC(=O)c1ccc([O-])cc1
IUPAC-namn sodium;4-ethoxycarbonylphenolate
Industry Type Clinical Testing
Molekylvikt (g/mol) 188.16
Kvantitet 10 mg
Formel vikt 188.0449 g/mol