missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Sodium Ferric EDDHA, Technical Grade, TRC
Specifications
Specifications
| Kemiskt namn eller material | Sodium Ferric EDDHA |
| CAS | 16455-61-1 |
| Molekylformel | C18 H16 N2 O6 . Fe . Na |
| Kvantitet | 100 mg |
| Synonym | Abreverd Fe-PM, Chelafer, Dissolvine Q, Dissolvine Q 15, Dissolvine Q 40 |
| InChI formel | InChI=1S/C18H20N2O6.Fe.Na/c21-13-7-3-1-5-11(13)15(17(23)24)19-9-10-20-16(18(25)26)12-6-2-4-8-14(12)22;;/h1-8,15-16,19-22H,9-10H2,(H,23,24)(H,25,26);;/q;+3;+1/p-4 |
| LEDER | [Na+].[Fe+3].[O-]C(=O)C(NCCNC(C(=O)[O-])c1ccccc1[O-])c2ccccc2[O-] |
| IUPAC-namn | sodium;2-[2-[[carboxylato-(2-oxidophenyl)methyl]amino]ethylamino]-2-(2-oxidophenyl)acetate;iron(3+) |
| Molekylvikt (g/mol) | 435.164 |
| Formel vikt | 435.026 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?