Läs mer
Sodium Fluoroacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Fluoroacetic acid sodium salt |
| CAS | 62-74-8 |
| Molekylformel | C2 H2 F O2 . Na |
| Kvantitet | 50 mg |
| Synonym | Fluoroacetic acid sodium, Acetic acid','2-fluoro-','sodium salt (1:1) (ACI), Acetic acid','fluoro-','sodium salt (8CI','9CI), 1080, Compound 1080, Fluoroacetic acid sodium salt, Fratol, Furatol, Ratbane 1080, SMFA, Sodium fluoroacetate, Sodium monofluoroacetate, Tenate |
| InChI formel | InChI=1S/C2H3FO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1 |
| LEDER | [Na+].[O-]C(=O)CF |
| IUPAC-namn | sodium;2-fluoroacetate |
| Molekylvikt (g/mol) | 100.02 |
| Formel vikt | 99.9937 |