missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide, 0.1M/l Solution, Scharlab Product Code.: 11398514

Sodium Hydroxide, 0.1M/l Solution, Scharlab

Product Code. p-7083585
1 L, 1 L
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
This item is not returnable. View return policy

Product Code. 11398514

Brand: Scharlab SO04431000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a titrant in volumetric analysis in analytical chemistry.

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5
  • Factor: 0.999–1.001, uncertainty ±0.001

This volumetric solution was checked by means of potentiometric methods using Scharlau's potassium hydrogen phthalate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow MoreShow Less
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Concentration 0.1mol/L
Concentration or Composition Notes 1mL = 0.00400g NaOH
Density 1.00g/cm3
Quantity 1 L
Formula Weight 40.00g/mol
Normality 0.1N
Traceability to NIST Traceable to NIST
Physical Form Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, 0.1M/l Solution, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.