missing translation for 'onlineSavingsMsg'
Läs mer

Natriumhydroxid, Scharlab

Product Code. 11388504
Change view
Click to view available options
Kvantitet:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
11388504 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11388504 Supplier Scharlab Supplier No. SO0425005P
 Begränsad artikel
Beräknad leverans: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1310-73-2
Molekylformel HNaO
InChI-nyckel HEMHJVSKTPXQMS-UHFFFAOYSA-M
Kvantitet 5 kg
LEDER [OH-].[Na+]
IUPAC-namn natriumhydroxid
Molekylvikt (g/mol) 40.00
Kvalitet Reagens
compliance-icons
Product Identifier
  • Natriumhydroxid, pellets, pro analysi, ACS, ISO
Signal Word
  • Fara
Hazard Category
  • Frätande eller irriterande verkan på hud Kategori 1A
Hazard Statement
  • H314-Orsakar allvarliga frätskador på hud och ögon.
Precautionary Statement
  • P260-Inandas inte damm/rök/gaser/dimma/ångor/ sprej.
  • P303+P361+P353-VID HUDKONTAKT (även håret): Ta omedelbart av alla nedstänkta kläder. Skölj huden med vatten/duscha.
  • P305+P351+P338-VID KONTAKT MED ÖGONEN: Skölj försiktigt med vatten i flera minuter. Ta ur eventuella kontaktlinser om det går lätt. Fortsätt att skölja.
  • P310-Kontakta genast GIFTINFORMATIONSCENTRALEN/läkare .
  • P405-Förvaras inlåst.
  • P501-Innehållet/behållaren lämnas till .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.