missing translation for 'onlineSavingsMsg'
Läs mer

Sodium Lauryl Sulfoacetate, TRC

Product Code. 30004949
Change view
Click to view available options
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30004949 100 mg
30007087 500 mg
30011272 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004949 Supplier Toronto Research Chemicals Supplier No. TRCS634303100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1847-58-1
Molekylformel C14H27O5SNa
Molekylvikt (g/mol) 330.42
Synonym Dodecyl sodium sulfoacetate, Lathanol, Lathanol LAL, Lathanol LAL 70, Miconate SLSA, Nacconol LAL, Nikkol LSA, Sodium lauryl sulfoacetate, Sulfo-dodecyl Ester Acetic Acid Sodium Salt (6CI), Sulfo-dodecyl Ester Acetic Acid S-Sodium Salt (7CI), Sulfo-1-dodecyl Ester Acetic Acid Sodium Salt (8CI,9CI);
IUPAC-namn sodium;2-dodecoxy-2-oxoethanesulfonate
LEDER [Na+].CCCCCCCCCCCCOC(=O)CS(=O)(=O)[O-]

Specifications

Kemiskt namn eller material Sodium Lauryl Sulfoacetate
Kvantitet 100 mg
InChI formel InChI=1S/C14H28O5S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-19-14(15)13-20(16,17)18;/h2-13H2,1H3,(H,16,17,18);/q;+1/p-1
Formel vikt 330.1477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.