Learn More
Sodium n-hexadecyl sulfate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium n-hexadecyl sulfate is used as an anionic surfactant.
Solubility
Soluble in water.
Notes
Incompatible with bases and strong oxidizing agents.
Sodium n-hexadecyl sulfate is used as an anionic surfactant.
Solubility
Soluble in water.
Notes
Incompatible with bases and strong oxidizing agents.
Specifications
Melting Point | 187°C to 192°C |
Quantity | 1 g |
Beilstein | 3578850 |
Solubility Information | Soluble in water. |
Formula Weight | 344.49 |
Percent Purity | 99% |
Chemical Name or Material | Sodium n-hexadecyl sulfate |