Läs mer
Sodium Nifurstyrenate, TRC
Produktkod. 30134116
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134116 50 mg
17927544 10 mg
17968534 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134116 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS61570050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54992-23-3
Molekylformel C13 H8 N O5 . Na
Molekylvikt (g/mol) 281.2
Synonym Nifurstyrenic Acid Sodium, Benzoic acid, 4-[2-(5-nitro-2-furanyl)ethenyl]-, sodium salt (1:1), Benzoic acid, 4-[2-(5-nitro-2-furanyl)ethenyl]-, sodium salt (9CI), Nifurstyrenate sodium, Sodium nifurstyrenate
IUPAC-namn sodium;4-[(E)-2-(5-nitrofuran-2-yl)ethenyl]benzoate
LEDER [Na+].[O-]C(=O)c1ccc(\C=C\c2oc(cc2)[N+](=O)[O-])cc1
Specifikationer
Kemiskt namn eller material Nifurstyrenic Acid Sodium
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H9NO5.Na/c15-13(16)10-4-1-9(2-5-10)3-6-11-7-8-12(19-11)14(17)18;/h1-8H,(H,15,16);/q;+1/p-1/b6-3+;
Formel vikt 281.03
Procent renhet >95%