missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrate, meets analytical spec. of FCC, E251, 99.0-100.5%, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | NaNO3 |
UN Number | 1498 |
Solubility Information | Soluble in water 1gm/1.1mL, Soluble in alcohol 1gm/125mL, Soluble in boling water 1gm/0.6mL, Soluble in boiling alcohol 1gm/52mL |
IUPAC Name | sodium;nitrate |
Formula Weight | 84.99g/mol |
Percent Purity | 99 to 100.5% |
Chemical Name or Material | Sodium Nitrate, Meets analytical specification of FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'