Läs mer
Sodium Nitrite-18O2 may contain up to 2% of 16O2, TRC
Produktkod. 30425169
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30425169 1 mg
1 options
Produktkod. 30425169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6480031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium Nitrite-18O2
CAS 123798-55-0
Molekylformel N 18O2 . Na
Kvantitet 1 mg
Synonym Nitrous-18O2 Acid Sodium Salt (10% 16O2)
InChI formel InChI=1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1/i2+2,3+2;
LEDER [Na+].[18O-]N=[18O]
Molekylvikt (g/mol) 72.995
Formel vikt 72.991