missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, (USP/FCC), MP Biomedicals
Click to view available options
Quantity:
500 g
1 kg
Specifications
Quantity | 500 g |
Formula Weight | 69 |
Grade | FCC/USP |
Chemical Name or Material | Sodium Nitrite |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'