Läs mer
Sodium nitroprusside, MedChemExpress
Produktkod. 16488366
Encompass_Preferred
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16488366 100 mg
16422176 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16488366 Leverantör Medchem Express Leverantörsnummer HYB0564
MedChemExpress Sodium nitroprusside (Ro 21-2498) is a potent vasodilator working through releasing NO spontaneously in blood.
Sodium nitroprusside (Ro 21-2498) is a potent vasodilator working through releasing NO spontaneously in blood.
Specifikationer
Kemiskt namn eller material Sodium nitroprusside
Fysisk form Solid
CAS 14402-89-2
Färg Reddish Brown
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C5FeN6Na2O
För användning med (applikation) COVID-19-immunoregulation
Synonym Ro 21-2498
Löslighetsinformation H2O : 100 mg/mL (381.80 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (127.25 mM; Need ultrasonic)
LEDER N#C[Fe-2](C#N)(C#N)(C#N)(N=O)C#N.[Na+].[Na+]
Molekylvikt (g/mol) 261.92
Formel vikt 261.92
Procent renhet 98.0%