Läs mer
Sodium Oleate, TRC
Produktkod. 30133091
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133091 10 g
30136590 50 g
30012609 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133091 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS96014010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 143-19-1
Molekylformel C18H33NaO2
Molekylvikt (g/mol) 304.44
IUPAC-namn sodium;(Z)-octadec-9-enoate
LEDER [Na+].CCCCCCCC\C=C/CCCCCCCC(=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium Oleate
Kvantitet 10 g
InChI formel InChI=1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;
Formel vikt 304.2378