Läs mer
Sodium Perfluoro-1-nonanesulfonate, TRC
Produktkod. 30003217
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003217 1 mg
30040985 5 mg
18082500 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003217 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0809451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98789-57-2
Molekylformel C9 F19 O3 S . Na
Molekylvikt (g/mol) 572.12
Synonym 1-Nonanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-, sodium salt (1:1), 1-Nonanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-, sodium salt (9CI), Sodium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-1-nonanesulfonate (1:1), Sodium perfluorononanesulfonate
IUPAC-namn sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane-1-sulfonate
LEDER [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material Perfluorononanesulfonic acid sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C9HF19O3S.Na/c10-1(11,2(12,13)4(16,17)6(20,21)8(24,25)26)3(14,15)5(18,19)7(22,23)9(27,28)32(29,30)31;/h(H,29,30,31);/q;+1/p-1
Formel vikt 571.9162
Procent renhet >95%