Läs mer
Sodium Perfluorodecanoate, TRC
Produktkod. 30411373
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30411373 500 mg
1 options
Produktkod. 30411373 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS665200500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluorodecanoic Acid Sodium
CAS 3830-45-3
Molekylformel C10 F19 O2 . Na
Kvantitet 500 mg
Synonym Decanoic acid','2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-','sodium salt (1:1), Decanoic acid','nonadecafluoro-','sodium salt (8CI,9CI), Sodium nonadecafluorodecanoate, Sodium perfluorocaprate, Sodium perfluorodecanoate, PFDoA','sodium salt
InChI formel InChI=1S/C10HF19O2.Na/c11-2(12,1(30)31)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29;/h(H,30,31);/q;+1/p-1
LEDER [Na+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
Molekylvikt (g/mol) 536.065
Formel vikt 535.9493