Läs mer
Sodium Perfluorohexanesulfonate, TRC
Produktkod. 17993235
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17993235 100 mg
17914255 50 mg
17972735 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17993235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS080865100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82382-12-5
Molekylformel C6 F13 O3 S . Na
Molekylvikt (g/mol) 422.0964
Synonym 1-Hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-, sodium salt (1:1), 1-Hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-, sodium salt (9CI), Sodium perfluorohexanesulfonate, PFHxS, Perfluorohexanesulfonic acid sodium, 1-Hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-, sodium salt (1:1), 1-Hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-, sodium salt (9CI), Sodium Perfluoro-1-hexanesulfonate, L-PFHxS
IUPAC-namn sodium;1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonate
LEDER [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material Sodium Perfluorohexanesulfonate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C6HF13O3S.Na/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22;/h(H,20,21,22);/q;+1/p-1
Formel vikt 421.9258
Procent renhet >95%