Läs mer
Sodium Permanganate Monohydrate, TRC
Produktkod. 30417806
Change view
Kvantitet:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417806 5 g
30417240 100 g
30415271 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6657005G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79048-36-5
Molekylformel H2MnNaO5
Molekylvikt (g/mol) 159.94
Synonym Permanganic Acid Sodium Salt Monohydrate
IUPAC-namn sodium;permanganate;hydrate
LEDER O.[Na+].[O-][Mn](=O)(=O)=O
Specifikationer
Kemiskt namn eller material Sodium Permanganate Monohydrate
Kvantitet 5 g
InChI formel InChI=1S/Mn.Na.H2O.4O/h;;1H2;;;;/q;+1;;;;;-1
Formel vikt 159.918