Läs mer
Sodium Phenylbutyrate, TRC
Produktkod. 30426960
Change view
Kvantitet:
50 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426960 5 g
30420648 10 g
30414636 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426960 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6670055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1716-12-7
Molekylformel C10 H11 O2 . Na
Molekylvikt (g/mol) 186.18
Synonym Sodium phenylbutyrate, Sodium 4-phenylbutanoate
IUPAC-namn sodium;4-phenylbutanoate
LEDER [Na+].[O-]C(=O)CCCc1ccccc1
Specifikationer
Kemiskt namn eller material Sodium phenylbutyrate
Kvantitet 5 g
InChI formel InChI=1S/C10H12O2.Na/c11-10(12)8-4-7-9-5-2-1-3-6-9;/h1-3,5-6H,4,7-8H2,(H,11,12);/q;+1/p-1
Formel vikt 186.0657