missing translation for 'onlineSavingsMsg'
Learn More

Sodium potassium (K2Na)-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™

Catalog Number 15453915
Click to view available options
Quantity:
5 g
25 g
100 g
MFCD08705348
MFCD08705348

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds
TRUSTED_SUSTAINABILITY
Color Black
Density 0.7 g/mL
Quantity 25 g
UN Number UN2813
Sensitivity Moisture sensitive
Concentration or Composition (by Analyte or Components) 35 to 40% alkali metal in silica gel
Physical Form Powder
Chemical Name or Material Sodium potassium (K2Na)-silica gel, Stage I
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'