Learn More
Sodium Rod, Diameter 2.5cm (Protective Liquid: Paraffin Oil), MilliporeSigma™
Product Code. 15854780
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity Unit Size
15854780 250 g 250g
1 options
Product Code. 15854780 Supplier Supelco Supplier No. 1062600250
 Restricted Item
Beräknad leverans: 19-11-2026
Specifications
CAS 7440-23-5
Appearance Silver solid
Quantity 250 g
Odor Odorless
Packaging Glass bottle
Form Solid
Molecular Formula Na
MDL Number MFCD00085307
UN Number UN 1428
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES [Na]
IUPAC Name sodium
Molecular Weight (g/mol) 22.99
PubChem CID 5360545
ChEBI CHEBI:26708
Formula Weight 22.99. (molar mass)
Chemical Name or Material Sodium