Learn More
Sodium Salicylate, For Analysis, EMSURE™, MilliporeSigma™
Product Code. 15884880
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15884880 1 kg Plastic Bottle 1kg
15874880 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15884880 Supplier Supelco Supplier No. 1066011000
 Restricted Item
Beräknad leverans: 25-11-2026
Recommended Storage
+5°C to +30°C.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Specifications
Melting Point 250°C (decomposition)
Color White
pH 8.7 to 9.3 (10g/L, H2O, 20°C)
Physical Form Crystals
Quantity 1 kg
Solubility Information 77g/L
Formula Weight 141.96
Percent Purity >99.0%
Grade Not Available
Packaging Plastic Bottle
Density 0.32g/3 (20°C)
Chemical Name or Material Sodium Salicylate