missing translation for 'onlineSavingsMsg'
Läs mer

Sodium Selenite, MP Biomedicals™

Produktkod. p-3547339 Handla allt MP Biomedicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
100 g
Förpackningsstorlek:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11492021 10 g 10g
11402031 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11492021 Leverantör MP Biomedicals Leverantörsnummer 0219474110

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Sodium Selenite is a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum.

  • Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine.
  • In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides.
  • Sodium selenite is included in various medium supplements for use in cell culture.
  • Sodium selenite has been utilized in studies of cell proliferation and cancer research.
  • It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays.
  • Sodium selenite can inhibit zinc finger protein/DNA interactions.
  • Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 10102-18-8
Molekylformel Na2O3Se
InChI-nyckel BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
PubChem CID 24934
ChEBI CHEBI:48843

Specifikationer

Kokpunkt Decomposition
Kemiskt namn eller material Sodium Selenite
Smältpunkt 1310°F (Decomposition)
Kvantitet 10 g
LEDER [O-][Se](=O)[O-].[Na+].[Na+]
IUPAC-namn disodium;selenite
Molekylvikt (g/mol) 172.948
Formel vikt 172.9
Procent renhet ∼98%
Kvalitet Cell Culture

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.