Läs mer
Sodium Stearyl Fumarate, TRC
Produktkod. 30427036
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427036 1 g
30420645 5 g
30413479 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427036 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6709501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4070-80-8
Molekylformel C22 H39 O4 . Na
Molekylvikt (g/mol) 390.53
Synonym Sodium stearyl fumarate, Sodium octadecyl (E)-butenedioate
IUPAC-namn sodium;(E)-4-octadecoxy-4-oxobut-2-enoate
LEDER [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C(=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium stearyl fumarate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C22H40O4.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20-26-22(25)19-18-21(23)24;/h18-19H,2-17,20H2,1H3,(H,23,24);/q;+1/p-1/b19-18+;
Formel vikt 390.2746
Procent renhet >95%