missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate USP, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 8028-06

Additional Details : CAS Number : 7757-82-6 Weight : 2.50000kg

Packaging Poly
Quantity 2.5kg

 View more versions of this product

Product Code. 15508464

  • 1425.01 SEK / 2.50kg


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Specifications

Chemical Identifiers

7757-82-6
142.036
24436
Na2O4S
PMZURENOXWZQFD-UHFFFAOYSA-L
CHEBI:32149

Specifications

Sodium Sulfate
Anhydrous
USP
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
Poly
142.04
7757-82-6
Na2O4S
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.036
CHEBI:32149
2.5kg
Videos