missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 3922-01

Additional Details : CAS Number : 7757-83-7 Weight : 0.64000kg

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Product Code. 15578184

  • 648.00 SEK / 500g


Estimated Shipment
21-01-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

7757-83-7
126.037
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
CHEBI:86477
[O-]S(=O)[O-].[Na+].[Na+]
Na2O3S
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
disodium;sulfite

Specifications

Sodium Sulfite
7757-83-7
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
[O-]S(=O)[O-].[Na+].[Na+]
126.037
CHEBI:86477
BAKER ANALYZED™ ACS Reagent
1L = 2.60kg
Anhydrous
Na2O3S
GEHJYWRUCIMESM-UHFFFAOYSA-L
disodium;sulfite
24437
126.04
Poly Bottle
500g
Videos
SDS
Documents