missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiophenolate, 90%, Thermo Scientific Chemicals

Catalog Number p-4258722
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
930-69-8
C6H5NaS
132.16
MFCD00066460
RZWQDAUIUBVCDD-UHFFFAOYSA-M
sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
4110039
sodium;benzenethiolate
[Na+].[S-]C1=CC=CC=C1
This item is not returnable. View return policy
930-69-8
C6H5NaS
132.16
MFCD00066460
RZWQDAUIUBVCDD-UHFFFAOYSA-M
sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
4110039
sodium;benzenethiolate
[Na+].[S-]C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color Beige to White
Physical Form Powder
Quantity 10 g
Assay Percent Range 88% min. (Iodimetry)
Linear Formula C6H5SNa
Solubility Information Solubility in water: soluble
Formula Weight 132.16
Percent Purity 90%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sodium thiophenolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium thiophenolate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'