Läs mer
Sodium Thiosulfate Hydrate, TRC
Specifications
Specifications
| Kemiskt namn eller material | Sodium Thiosulfate hydrate |
| CAS | 128114-51-2 |
| Molekylformel | 2Na . O3 S2 . x(H2 O) |
| Kvantitet | 10 mg |
| Synonym | Thiosulfuric Acid (H2S2O3) Disodium Salt Hydrate, Thiosulfuric Acid Disodium Salt Hydrate, Disodium Thiosulfate Hydrate, Disodium Thiosulphate Hydrate, Hypo Sodium Thiosulfate (Na2S2O3) Hydrate |
| InChI formel | InChI=1S/2Na.H2O3S2.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2 |
| LEDER | O.[Na+].[Na+].[O-]S(=O)(=S)[O-] |
| IUPAC-namn | disodium;dioxido-oxo-thioxo-$l^{6}-sulfane;hydrate |
| Molekylvikt (g/mol) | 176.13 |